Learn More
IL-2 Mouse anti-Human, CoraLite 488, Clone: 2F11C5, Proteintech
Mouse Monoclonal Antibody
Merk: Proteintech CL488-60306-100UL
This item is not returnable.
View return policy
Beschrijving
The IL-2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IL-2. This antibody recognizes human antigen. The IL-2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specificatie
IL-2 | |
Monoclonal | |
CoraLite 488 | |
100 μL | |
3558 | |
IgG2a |
Immunofluorescence | |
2F11C5 | |
Mouse | |
Primary | |
Human |