missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated GAPDH Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Merk: Proteintech CL488-60004-20UL
This item is not returnable.
View return policy
Beschrijving
The GAPDH antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GAPDH. This antibody recognizes human, mouse, rat, zebrafish, yeast, plant antigen. The GAPDH antibody has been validated for the following applications: WB, FC (Intra) analysis. This antibody is coralite™ plus 488.Specificatie
GAPDH | |
Monoclonal | |
1000 μg/mL | |
G3PD; GAPD; MGC88685 | |
Recombinant Protein | |
20 μL | |
Primary | |
Human, Mouse, Rat, Zebrafish, Yeast, Plant | |
36 kDa | |
Liquid |
Western Blot, Flow Cytometry (Intracellular Staining) | |
1E6D9 | |
CoraLite Plus 488 | |
Mouse | |
Protein A purification | |
RUO | |
2597 | |
Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
Conjugated Antibody | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction