Learn More
Macherey-Nagel™ Cellulose Mn
Merk: Macherey-Nagel™ 816250.1
This item is not returnable.
View return policy
Specificatie
1 kg | |
Cellulose Mn | |
Cellulose |
Product Content Correction
Product Title
Macherey-Nagel™ Cellulose Mn > Quantity: 1kg