missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Atlantic Blue™ Anti-Mouse CD4 (RM4-4), Proteintech
Rat Monoclonal Antibody
Merk: Proteintech AB-65199-100UG
This item is not returnable.
View return policy
Beschrijving
The CD4 antibody from Proteintech is a rat monoclonal antibody generated with balb/c mouse thymocytes of mouse CD4. This antibody recognizes mouse antigen. The CD4 antibody has been validated for the following applications: FC analysis. This antibody is atlantic blue™.Specificatie
CD4 | |
Monoclonal | |
0.5 mg/mL | |
Rat | |
Affinity purification | |
RUO | |
12504 | |
Store at 2-8°C. Avoid exposure to light. Stable for one year after shipment. | |
Liquid |
Flow Cytometry | |
RM4-4 | |
Atlantic Blue | |
BALB/c mouse thymocytes | |
100 μg | |
Primary | |
Mouse | |
Flow Antibody | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction