Learn More
Uvex™ 9174 Skylite Safety Glasses
Skylite safety glasses are extremely light, combining a high level of comfort with effective eye protection.
Specificatie
Lenstype | Ultradura |
---|---|
Model | 9174 Skylite |
Lens Material | Polycarbonate |
Beschrijving
- Safety glasses are extremely lightweight, combining comfort with effective eye protection
- Certified in line with EN 166 (personal eye protection)
- Ultradura lens coating is extremely scratch-resistant on the interior and exterior
- Integrated brow and cheek guards for increased wearer protection
- Spectacles protect against liquid droplets/splashes, and large dust particles
- Flexible, closed-pore soft components of frame adapt to individual face contours
- Easy cleaning process due to separate lens and frame concept
- 5-stage side arm length adjustment ensures optimum fit
Specificatie
Ultradura | |
9174 Skylite | |
Polycarbonate |