Learn More
Uvex™ 9174 Skylite Safety Glasses
Skylite safety glasses are extremely light, combining a high level of comfort with effective eye protection.
Specificatie
Lenstype Ultradura
Model 9174 Skylite
Lens Material Polycarbonate
Meer specificaties weergeven
Beschrijving
Beschrijving
  • Safety glasses are extremely lightweight, combining comfort with effective eye protection
  • Certified in line with EN 166 (personal eye protection)
  • Ultradura lens coating is extremely scratch-resistant on the interior and exterior
  • Integrated brow and cheek guards for increased wearer protection
  • Spectacles protect against liquid droplets/splashes, and large dust particles
  • Flexible, closed-pore soft components of frame adapt to individual face contours
  • Easy cleaning process due to separate lens and frame concept
  • 5-stage side arm length adjustment ensures optimum fit
Specificatie
Specificatie
Ultradura
9174 Skylite
Polycarbonate